Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC286149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093190.1 | - | 1 | 159 |
Gene Symbol: LOC105375650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132987.1 | 1 | 159 | |||
GenBank mRNA | BC128222.1 | - | 2 | 768 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC128221.1 | - | 1 | 860 | Chr. 8 - 95072725 - 95073621 [-] on Build GRCh38 |