Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306215.1 | NP_001293144.1 | 15 | 5010 | ||
NM_178835.4 | NP_849157.2 | 15 | 5019 | |||
XM_011531634.2 | XP_011529936.1 | 14 | 5024 | |||
XM_011531635.2 | XP_011529937.1 | 15 | 10511 | |||
XM_011531645.2 | XP_011529947.1 | 10 | 2762 | |||
XM_017007768.1 | XP_016863257.1 | 14 | 5019 | |||
XM_017007769.1 | XP_016863258.1 | 14 | 11270 | |||
XM_017007770.1 | XP_016863259.1 | 13 | 4791 | |||
XM_017007777.1 | XP_016863266.1 | 11 | 2761 | |||
GenBank mRNA | AK094136.1 | - | 10 | 2299 | ||
AK131430.1 | - | 1 | 2817 | |||
BC033248.1 | - | 1 | 173 |