Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIVEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038260.1 | 3 | 1445 | |||
NR_038261.1 | 4 | 1263 | ||||
GenBank mRNA | BC015242.1 | - | 1 | 526 | ||
BC040942.1 | - | 3 | 1289 | |||
BC046120.1 | - | 4 | 1263 |