Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100131289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038929.1 | 3 | 771 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK130456.1 | - | 2 | 1376 | Chr. 6 - 27761503 - 27763267 [+] on Build GRCh38 |