Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270875.1 | NP_001257804.1 | 7 | 4700 | ||
NM_001270876.1 | NP_001257805.1 | 6 | 4373 | |||
NM_022756.5 | NP_073593.2 | 8 | 4730 | |||
NR_073090.1 | 9 | 4788 | ||||
NR_073091.1 | 8 | 4758 | ||||
NR_073092.1 | 9 | 4819 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF172817.1 | - | 1 | 79 | Chr. 1 - 37487118 - 37490126 [-] on Build GRCh38 |