Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100505564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055529.1 | - | 1 | 287 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA559788.1 | - | 1 | 287 | Chr. 16 - 10940719 - 10941427 [+] on Build GRCh38 |