Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110818.1 | 1 | 1053 | |||
GenBank mRNA | BC069263.1 | - | 1 | 1053 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000574885.1 | - | 1 | 1053 | Chr. 17 - 2962248 - 2965895 [-] on Build GRCh38 |