Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC002421.2 | - | 1 | 1426 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA219844.1 | - | 1 | 17 | Chr. 8 - 24912741 - 24913308 [-] on Build GRCh38 |