Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005248250.3 | XP_005248307.2 | 11 | 3833 | ||
XM_011513966.2 | XP_011512268.2 | 12 | 3968 | |||
GenBank mRNA | AK022694.1 | - | 1 | 411 | ||
AK054651.1 | - | 3 | 2383 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU125587.1 | - | 1 | 411 | Chr. 5 - 34952189 - 34952908 [+] on Build GRCh38 |