Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100132764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131376.1 | - | 1 | 3823 |
Gene Symbol: CADM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047021.1 | - | 9 | 1511 | 5248100 |