Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100131180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124533.1 | - | 1 | 3697 |
Gene Symbol: APTX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014836.1 | XP_016870325.1 | 8 | 991 |