Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNU13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028811.1 | XP_016884300.1 | 3 | 822 | ||
GenBank mRNA | AK124020.1 | - | 1 | 1065 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG822290.1 | - | 1 | 587 | Chr. 22 - 41675038 - 41688328 [-] on Build GRCh38 |