Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092477.1 | - | 1 | 1034 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF980627.1 | - | 409 | Chr. - - [-] on Build |