Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833410.1 | - | 1 | 907 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA016019.1 | - | 1 | 475 | Chr. 7 - 134443665 - 134444215 [-] on Build GRCh38 |