Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6PR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057556.1 | - | 1 | 1808 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN286038.1 | - | 1 | 107 | Chr. 12 - 8947613 - 8948266 [-] on Build GRCh38 |