Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLJ44715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126671.1 | - | 1 | 701 |
Gene Symbol: SEC24C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA419040.1 | - | 1 | 32 |