Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLJ44715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126671.1 | - | 1 | 71 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI833237.1 | - | 1 | 106 | Chr. 10 - 73772157 - 73772897 [-] on Build GRCh38 |