Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF086515.1 | - | 1 | 52 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | W95185.1 | - | 52 | Chr. - - [-] on Build |