Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542750.2 | XP_011541052.1 | 11 | 1768 | ||
GenBank mRNA | AK124235.1 | - | 1 | 484 |