Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U95646.1 | - | 49 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK000561.1 | - | 262 | Chr. - - [-] on Build |