Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721996.3 | XP_006722059.3 | 1 | 4525 | ||
XM_006721997.3 | XP_006722060.3 | 1 | 4525 | |||
XM_006722000.3 | XP_006722063.3 | 1 | 4525 | |||
XM_017024906.1 | XP_016880395.1 | 1 | 4525 | |||
XM_017024907.1 | XP_016880396.1 | 1 | 4525 | |||
XM_017024908.1 | XP_016880397.1 | 1 | 4525 | |||
GenBank mRNA | BC035105.1 | - | 1 | 3419 | ||
BC051468.1 | - | 1 | 3414 |