Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023988.1 | XP_016879477.1 | 5 | 501 | ||
XM_017023989.1 | XP_016879478.1 | 5 | 467 | |||
XM_017023990.1 | XP_016879479.1 | 5 | 492 | |||
XM_017023991.1 | XP_016879480.1 | 5 | 501 | |||
XM_017023992.1 | XP_016879481.1 | 5 | 502 | |||
XM_017023993.1 | XP_016879482.1 | 5 | 501 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK098202.1 | - | 1 | 2206 | Chr. 17 - 44102377 - 44105335 [-] on Build GRCh38 |