Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928068 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111907.1 | 2 | 524 | |||
GenBank mRNA | AK095945.1 | - | 2 | 1549 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | EL946683.1 | - | 1 | 421 | Chr. 1 - 244969448 - 244969900 [-] on Build GRCh38 |