Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713592.3 | XP_006713655.1 | 19 | 15042 | ||
XM_017006210.1 | XP_016861699.1 | 18 | 14916 | |||
XM_017006211.1 | XP_016861700.1 | 20 | 15067 | |||
XM_017006212.1 | XP_016861701.1 | 19 | 14736 | |||
XM_017006213.1 | XP_016861702.1 | 19 | 15094 | |||
XM_017006214.1 | XP_016861703.1 | 18 | 14362 | |||
XM_017006215.1 | XP_016861704.1 | 14 | 13391 | |||
XM_017006216.1 | XP_016861705.1 | 13 | 13341 | |||
XM_017006217.1 | XP_016861706.1 | 15 | 13438 | |||
XM_017006218.1 | XP_016861707.1 | 13 | 13241 | |||
XM_017006220.1 | XP_016861709.1 | 15 | 13406 | |||
XM_017006221.1 | XP_016861710.1 | 13 | 13352 | |||
XM_017006222.1 | XP_016861711.1 | 14 | 13400 | |||
GenBank mRNA | AK000957.1 | - | 1 | 230 |