Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011539270.2 | XP_011537572.1 | 7 | 7038 | ||
XM_011539273.2 | XP_011537575.1 | 5 | 5455 | |||
GenBank mRNA | AK094859.1 | - | 1 | 1262 | ||
AK097894.1 | - | 1 | 1696 | |||
AK126230.1 | - | 1 | 2600 | |||
AL831850.1 | - | 1 | 1617 |