Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006710946.3 | XP_006711009.1 | 5 | 3782 | ||
GenBank mRNA | AF085890.1 | - | 1 | 384 | ||
AK075186.1 | - | 1 | 969 |