Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104367.2 | 14 | 2158 | |||
NR_104368.2 | 12 | 1352 | ||||
GenBank mRNA | AY331564.1 | - | 14 | 2029 | ||
AY333779.1 | - | 12 | 1223 | |||
AY353056.1 | - | 3 | 347 | |||
BX641062.1 | - | 1 | 1553 |