Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021569.1 | XP_016877058.1 | 1 | 2391 | ||
XM_017021570.1 | XP_016877059.1 | 1 | 2391 | |||
XM_017021571.1 | XP_016877060.1 | 1 | 2391 | |||
XM_017021572.1 | XP_016877061.1 | 1 | 2391 | |||
XM_017021574.1 | XP_016877063.1 | 1 | 2391 | |||
XM_017021575.1 | XP_016877064.1 | 1 | 2391 | |||
XM_017021577.1 | XP_016877066.1 | 1 | 2391 | |||
GenBank mRNA | BC045740.1 | - | 1 | 1278 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK023121.1 | - | 1 | 2392 | Chr. 14 - 23005503 - 23008236 [-] on Build GRCh38 |