Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFT2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145169.2 | NP_660152.1 | 8 | 932 | ||
NR_130112.1 | 9 | 955 | ||||
NR_130113.1 | 7 | 860 | ||||
NR_130114.1 | 7 | 851 | ||||
GenBank mRNA | AF041429.1 | - | 8 | 866 | ||
AK090795.1 | - | 1 | 1839 | |||
AL833217.1 | - | 2 | 3263 | |||
BC028388.1 | - | 8 | 2550 |