Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023106.1 | XP_016878595.1 | 3 | 2139 | ||
GenBank mRNA | AK123825.1 | - | 1 | 1455 |