Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039960.2 | NP_001035049.1 | 25 | 8943 | ||
NM_001258401.2 | NP_001245330.1 | 25 | 8889 | |||
XM_011539010.1 | XP_011537312.1 | 26 | 8998 | |||
XM_011539011.1 | XP_011537313.1 | 26 | 8943 | |||
XM_011539014.2 | XP_011537316.1 | 16 | 7596 | |||
XM_017020240.1 | XP_016875729.1 | 26 | 9025 | |||
XM_017020242.1 | XP_016875731.1 | 26 | 9012 | |||
GenBank mRNA | AK124576.1 | - | 1 | 2141 | ||
AL137310.1 | - | 1 | 897 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA194498.1 | - | 1 | 566 | Chr. 12 - 51512356 - 51512965 [+] on Build GRCh38 |