Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42EP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270436.1 | NP_001257365.1 | 2 | 4496 | ||
NM_001270437.1 | NP_001257366.1 | 2 | 4452 | |||
NM_001270438.1 | NP_001257367.1 | 2 | 4428 | |||
NM_006449.4 | NP_006440.2 | 2 | 5087 | |||
XM_011532482.2 | XP_011530784.1 | 2 | 4893 | |||
GenBank mRNA | AK055915.1 | - | 1 | 1626 |