Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012409.3 | NP_001012409.1 | 8 | 2556 | ||
NM_001012411.3 | NP_001012411.1 | 8 | 1800 | |||
NM_001012413.3 | NP_001012413.1 | 7 | 1749 | |||
NM_001199251.2 | NP_001186180.1 | 8 | 2667 | |||
NM_001199253.2 | NP_001186182.1 | 8 | 1911 | |||
NM_001199255.2 | NP_001186184.1 | 7 | 1860 | |||
NR_131179.1 | 7 | 1603 | ||||
NR_131180.1 | 6 | 1552 | ||||
GenBank mRNA | AK024292.1 | - | 1 | 742 | ||
BX648516.1 | - | 7 | 1834 |