Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKBKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273490.2 | XP_005273547.1 | 21 | 2991 | ||
XM_005273491.4 | XP_005273548.1 | 20 | 2891 | |||
XM_005273493.3 | XP_005273550.1 | 18 | 2687 | |||
XM_005273495.2 | XP_005273552.1 | 20 | 2732 | |||
XM_005273496.3 | XP_005273553.1 | 19 | 2752 | |||
XM_005273498.3 | XP_005273555.1 | 17 | 2561 | |||
XM_011544517.2 | XP_011542819.1 | 21 | 2989 | |||
XM_011544518.2 | XP_011542820.1 | 20 | 2841 | |||
XM_011544519.2 | XP_011542821.1 | 20 | 2859 | |||
XM_011544520.2 | XP_011542822.1 | 18 | 2706 | |||
XM_011544521.2 | XP_011542823.1 | 19 | 2819 | |||
XM_011544522.2 | XP_011542824.1 | 18 | 2667 | |||
XM_017013396.1 | XP_016868885.1 | 20 | 2944 | |||
GenBank mRNA | AK095687.1 | - | 1 | 477 |