Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKBKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091090.1 | - | 1 | 461 | ||
AK093537.1 | - | 1 | 782 | |||
AK095687.1 | - | 1 | 1480 |