Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERINC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174072.2 | NP_001167543.1 | 12 | 6140 | ||
XM_011543304.1 | XP_011541606.1 | 12 | 6122 | |||
XM_017009323.1 | XP_016864812.1 | 12 | 6103 | |||
XM_017009324.1 | XP_016864813.1 | 12 | 6122 | |||
XM_017009325.1 | XP_016864814.1 | 10 | 5758 | |||
XM_017009326.1 | XP_016864815.1 | 10 | 5741 | |||
GenBank mRNA | AK026295.1 | - | 1 | 1099 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU561394.1 | - | 1 | 614 | Chr. 5 - 80138940 - 80139682 [-] on Build GRCh38 |