Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015348.1 | XP_016870837.1 | 17 | 2908 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK023463.1 | - | 1 | 702 | Chr. 9 - 127215249 - 127217341 [+] on Build GRCh38 |