Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190728.1 | NP_001177657.1 | 17 | 1962 | ||
NM_001190729.1 | NP_001177658.1 | 18 | 2225 | |||
NM_001322320.1 | NP_001309249.1 | 18 | 2188 | |||
NM_001322323.1 | NP_001309252.1 | 17 | 2135 | |||
NM_001322325.1 | NP_001309254.1 | 20 | 2435 | |||
XM_006717328.3 | XP_006717391.2 | 21 | 2409 | |||
XM_011519225.1 | XP_011517527.1 | 20 | 2355 | |||
XM_011519226.1 | XP_011517528.1 | 20 | 2334 | |||
XM_011519227.1 | XP_011517529.1 | 19 | 2307 | |||
XM_011519229.1 | XP_011517531.1 | 21 | 2415 | |||
XM_011519230.2 | XP_011517532.1 | 21 | 2409 | |||
XM_011519231.1 | XP_011517533.1 | 18 | 2148 | |||
XM_011519232.1 | XP_011517534.1 | 18 | 2145 | |||
XM_011519234.2 | XP_011517536.1 | 20 | 2492 | |||
XM_011519235.2 | XP_011517537.1 | 21 | 2583 | |||
XM_017015343.1 | XP_016870832.1 | 21 | 2402 | |||
XM_017015344.1 | XP_016870833.1 | 18 | 2175 | |||
XM_017015348.1 | XP_016870837.1 | 17 | 4279 | |||
XM_017015349.1 | XP_016870838.1 | 17 | 2043 | |||
GenBank mRNA | AK299149.1 | - | 18 | 2166 | ||
CB162049.1 | - | 1 | 58 | |||
DB521440.1 | - | 2 | 433 |