Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAALC-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109954.1 | 2 | 443 | |||
GenBank mRNA | AK125733.1 | - | 7 | 1623 | ||
AK311189.1 | - | 2 | 443 | |||
DA226722.1 | - | 2 | 444 | |||
DA333188.1 | - | 2 | 331 | |||
DT217610.1 | - | 2 | 330 |