Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANSL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003431.1 | XP_016858920.1 | 1 | 2443 | ||
GenBank mRNA | AK057810.1 | - | 1 | 101 |