Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000855.2 | NP_000846.1 | 8 | 5036 | ||
NM_001256424.1 | NP_001243353.1 | 9 | 5129 | |||
GenBank mRNA | AK098142.1 | - | 1 | 1687 |