Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZRN3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046681.1 | 2 | 1939 | |||
GenBank mRNA | AK126423.1 | - | 1 | 2543 |