Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535703.2 | XP_011534005.1 | 8 | 7270 | ||
XM_011535704.2 | XP_011534006.1 | 8 | 7148 | |||
XM_011535706.1 | XP_011534008.1 | 7 | 7114 | |||
XM_011535709.2 | XP_011534011.1 | 4 | 7304 | |||
XM_011535710.2 | XP_011534012.1 | 6 | 6981 | |||
XM_017010677.1 | XP_016866166.1 | 8 | 7439 | |||
XM_017010678.1 | XP_016866167.1 | 8 | 7352 | |||
XM_017010679.1 | XP_016866168.1 | 8 | 7250 | |||
XM_017010680.1 | XP_016866169.1 | 8 | 7318 | |||
XM_017010681.1 | XP_016866170.1 | 8 | 7228 | |||
XM_017010682.1 | XP_016866171.1 | 8 | 7453 | |||
XM_017010683.1 | XP_016866172.1 | 7 | 7111 | |||
XM_017010685.1 | XP_016866174.1 | 7 | 7238 | |||
XM_017010686.1 | XP_016866175.1 | 7 | 7223 | |||
XM_017010687.1 | XP_016866176.1 | 7 | 7149 | |||
GenBank mRNA | AK128420.1 | - | 1 | 897 |