Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSLTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282186.1 | NP_001269115.1 | 2 | 2178 | ||
NM_001282187.1 | NP_001269116.1 | 4 | 2348 | |||
NM_001282188.1 | NP_001269117.1 | 4 | 2325 | |||
NM_006639.3 | NP_006630.1 | 3 | 2265 | |||
GenBank mRNA | DQ131803.1 | - | 1 | 822 |
Set Membership: |
G-protein coupled receptor |