Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC022394.1 | - | 1 | 1204 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA301100.1 | - | 1 | 156 | Chr. 19 - 21398214 - 21398470 [+] on Build GRCh38 |