Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGGT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005272020.3 | XP_005272077.2 | 10 | 7449 | ||
GenBank mRNA | AK096154.1 | - | 1 | 236 |