Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGGT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005272020.3 | XP_005272077.2 | 10 | 9060 | ||
XM_011543490.2 | XP_011541792.1 | 10 | 1492 | |||
XM_011543492.2 | XP_011541794.1 | 10 | 1489 | |||
GenBank mRNA | AK096154.1 | - | 1 | 1853 |