Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003403.4 | NP_003394.1 | 5 | 2684 | ||
GenBank mRNA | AF147416.1 | - | 1 | 182 | ||
DA234524.1 | - | 1 | 113 |
Gene Symbol: YY1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033658.1 | 1 | 1542 | |||
GenBank mRNA | BC042175.1 | - | 1 | 1539 |