Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028290.1 | XP_016883779.1 | 1 | 2472 | ||
GenBank mRNA | AY216265.1 | - | 1 | 1199 |