Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012736.1 | XP_016868225.1 | 1 | 819 | ||
GenBank mRNA | AY129022.1 | - | 1 | 48 |